Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Actual product may vary
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Line
5045-0003BlueNalgene
354755WhiteNunc
354879BlueNunc
354968RedNunc
355018GreenNunc
355077YellowNunc
375868BrownNunc
375884PinkNunc
375906GrayNunc
375922PurpleNunc
5045-0000WhiteNalgene
5045-0002YellowNalgene
5045-0004GreenNalgene
5045-0005RedNalgene
Catalog number 5045-0003
Price (MYR)
580.00
CS
Add to cart
Color:
Blue
Product Line:
Nalgene
Price (MYR)
580.00
CS
Add to cart
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
DescriptionNalgene™ Color coder
ColorBlue
MaterialPolycarbonate
Product LineNalgene
Quantity100/Pk., 500/Cs.
TypeVial Closure
Unit SizeCS