Cryoware Labels
Cryoware Labels
Actual product may vary
Cryoware Labels
Cryoware Labels
Cryoware Labels
Cryoware Labels
Cryoware Labels
Easily and permanently identify samples and storage boxes with Thermo Scientific™ Cryoware Labels.
Have Questions?
Catalog NumberDimensions (LxW)
5040-000225 x 50 mm
Catalog number 5040-0002
Price (MYR)
3,989.00
CS
Add to cart
Dimensions (LxW):
25 x 50 mm
Price (MYR)
3,989.00
CS
Add to cart
Easily and permanently identify samples and storage boxes with Thermo Scientific™ Cryoware Labels.
  • 25 x 50mm cloth labels
  • Accept markers or ballpoint pen
  • Specifically designed for use at ultra-low temperatures
  • Adhere to plastic and cardboard without shrinking or peeling
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Specifications
AutoclavableNot Autoclavable
Adhesive TypeCold-temperature
ColorWhite
MaterialNylon Cloth
SterilityNon-sterile
Latex FreeYes
Dimensions (LxW)25 x 50 mm
Voltage0/1 V
Unit SizeCS