Heratherm™ Compact Microbiological Incubators
Heratherm™ Compact Microbiological Incubators
Actual product may vary
Heratherm™ Compact Microbiological Incubators
Save space with our compact microbiological incubator, which is ideal for a personalized workspace or crowded labs with small sample volumes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug Type
50125882EU, UK, Australia
50125590Nema 5-15 US
Catalog number 50125882
HSN 84198990
Price (MYR)
7,024.00
EA
Add to cart
Plug Type:
EU, UK, Australia
Price (MYR)
7,024.00
EA
Add to cart
Save space with the Thermo Scientific™ Heratherm™ Compact Microbiological Incubator. The extremely small footprint makes it ideal for a personalized workspace or crowded labs with small sample volumes. The most compact unit of the Heratherm microbiological incubators family has an 18L capacity, and operates in temperatures at or below ambient.
Features:
  • Minimal benchtop space for space restricted lab areas
  • Easy-to-use interface
  • Internal light and window in door facilitates sample observation
Temperature Security:
  • Temperatures at or below ambient: 17°C to 50°C
  • Temperature uniformity of ±1.2°C — measured at 37°C
  • Temperature stability of ± 0.2°C — measured at 37°C
  • High temperature accuracy
Ordering Information:
Availability may vary by country.
Warranty: 2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)0.65 cu. ft.
Capacity (English) Shelf4.4 lb.
Certifications/CompliancecCSAus, CE
Controller TypeLED display
Capacity (Metric)18 L
Convection TypeMechanical
No. of Shelves2 supplied/ 3 max.
Plug TypeEU, UK, Australia
Temperature Range (Metric)17°C to 50°C
TypeMicrobiological Incubator
Temperature Stability±0.2°C
Temperature Uniformity±1.2°C
Dimensions (D x W x H) Interior11.4 x 7.1 x 12.2 in. (29 x 18 x 31 cm)
Dimensions (L x W x H)18.5 x 10.2 x 16.3 in. (47 x 26 x 41.5 cm)
Length (English)18.5 in.
Length (Metric)47 cm
Width (English)10.2 in.
Width (English) Interior7.1 in.
Width (Metric)26 cm
Width (Metric) Interior18 cm
Height (English)16.3 in.
Height (English) Interior12.2 in.
Height (Metric)41.5 cm
Height (Metric) Interior31 cm
Depth (English) Interior11.4 in.
Depth (Metric) Interior29 cm
Weight (English)15.9 lb.
Weight (Metric)7.2 kg
Electrical Requirements100/240 V, 50/60 Hz
Voltage100/240 V
Wattage45 w
Amperage0.45 to 0.85 A