13mm Clear Glass Screw Thread Vials
13mm Clear Glass Screw Thread Vials
Actual product may vary
13mm Clear Glass Screw Thread Vials
13mm Clear Glass Screw Thread Vials
13mm Clear Glass Screw Thread Vials
These 13mm vials are manufactured from high-quality glass and fit a variety of autosamplers.
Have Questions?
Catalog NumberVolume (Metric)TypeBase StyleMarking Spot
5-SV5.0 mLScrew Thread VialRound BottomNo
Catalog number 5-SV
Price (MYR)
-
Volume (Metric):
5.0 mL
Type:
Screw Thread Vial
Base Style:
Round Bottom
Marking Spot:
No
These 13mm vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) glass and feature a 13-425 thread finish, they are available in Thermo Scientific™ Premiere™, National™ and Chromacol™ brands.
Choose the right vial for your needs
Get maximum sample extraction without need for separate inserts by using Microsampling and High Recovery Vials
Identify sample and volume with graduated, write-on patch provided on certain vials
Obtain maximum recovery of critical polar, labile or OH-interacting compounds using silanized (deactivated) glass (optional)
Polyspring inserts are self-aligning and provide a cushion against needle contact.
Autosampler compatibility*
Shimadzu™
Spark Holland™
Varian™
VWR™ (Merck)/Hitachi™
Waters™ (Wisp 48 Position Carousel)
*See the Columns and Consumables Catalog for autosampler compatibility
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description13-425 Screw Thread Vials
Diameter (Metric) Opening13 mm
Diameter (Metric) Inner13 mm
ColorClear
Diameter (Metric) Outer13 mm
MaterialGlass
Volume (Metric)5.0 mL
No. per Pack125
Volume (Metric) Residual<500 μL
Base StyleRound Bottom
Volume (Metric) Total5 mL
Closure TypeScrew Cap
Height (Metric)65 mm
TypeScrew Thread Vial
Closure Size13-425
Marking SpotNo
Volume (Metric) Usable4.5 mL
Unit SizeBox