Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
Actual product may vary
Finnpipette™ F2 Good Laboratory Pipetting (GLP) Kits
Enjoy the convenience of having a variety of Finnpipette™ F2 single channels packaged together, in a range of volumes, with compatible Finntip pipette tips and a pipette stand to support good laboratory pipetting in your lab
Have Questions?
Catalog NumberVolume (Metric)
470088510 to 10,000 μL
Catalog number 4700885
Price (MYR)
5,381.00
EA
Add to cart
Volume (Metric):
10 to 10,000 μL
Price (MYR)
5,381.00
EA
Add to cart
Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F2 GLP (Good Laboratory Pipetting) Kits provide durable, high-performance Finnpipette F2 pipettes which offer maximum reliability and comfort. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Features:
  • Fully autoclavable for protection
  • Tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light
  • Easy maintenance
  • Color coded together with Finntips for easier recognition of compatible tips
Kit 1 includes:
  • Three Thermo Scientific Finnpipette F2 Pipettes from 1 to 1000 μL:
    • 1-10 μL micro
    • 10-100 μL
    • 100-1000 μL
  • Flex tips: 10 micro: 1 x 96; 200: 1 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack
Kit 2 includes:
  • Four Thermo Scientific Finnpipette F2 Pipettes from 0.2 to 1000 μL:
    • 0.2-2 μL
    • 2-20 μL univ
    • 20-200 μL
    • 100-1000 μL
  • Flex tips: 10 micro: 1 x 96; 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack
Kit 3 includes:
  • Three Thermo Scientific Finnpipette F2 Pipettes from 10 to 10,000 μL:
    • 10-100 μL
    • 100-1000 μL
    • 1000-10,000 μL
  • Flex tips: 200: 1 x 96; 1000: 1 x 96; FT 10 mL: 1 x 24
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack
Kit 4 includes:
  • Three Thermo Scientific Finnpipette F2 Pipettes from 20 to 1000 μL:
    • 2-20 μL
    • 20-200 μL
    • 100-1000 μL 
  • Flex tips: 200: 2 x 96; 1000: 1 x 96
  • F-stand
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack
Ordering Information:
The kit contains samples of Finntip Flex, the most advanced traditional tip offering very low attachment and ejection forces. The Good Laboratory Pipetting Guide contains helpful tips for pipetting including pipetting techniques, ergonomic considerations and how to select tips for best performance.
Warranty: 5 years with web registration.
Specifications
Compatible TipsFlex Tips: 200, 1000 μL, 10 mL
DescriptionFinnpipette F2 GLP Kit 3: 10-100μL, 100-1000μL and 1-10mL
AutoclavableAutoclavable
ColorAssorted
ModelF2
Number of Channels1
Product LineFinnpipette F2
TypeGLP Kit
Volume (Metric)10 to 10,000 μL
Unit SizeEA