Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Actual product may vary
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Finnpipette™ F3 Variable Volume Single Channel Pipettes
Support every day pipetting in comfort and with ideal function using these manual variable volume pipettes.
Have Questions?
Catalog NumberVolume (Metric)Color Code
46400805 to 50 μLTurquoise
Catalog number 4640080
HSN 84798999
Price (MYR)
991.00
EA
Add to cart
Volume (Metric):
5 to 50 μL
Color Code:
Turquoise
Price (MYR)
991.00
EA
Add to cart
Thermo Scientific™ Finnpipette™ F3 Variable Volume Single Channel Pipettes are comfortable to use, and provide ideal functionality in every day pipetting. The Finnpipette F3 pipette features a wide, color-coded supportive finger rest and ergonomic handle design. Finnpipette F3 pipettes are available in single and multichannel models.
Ergonomic Comfort
  • Wide color-coded supportive finger rest for identification support and ergonomics
  • Designed handle softly contours the shape of your hand
  • Large Ergovisio Display for volume recognition
Ideal Function
  • Durable handle material
  • Double-action pipetting button; rotating upper part prevents accidental volume alteration
  • Advanced volume gearing mechanism for enhanced precision and accuracy
  • Super blow-out for efficient delivery of micro-size drops in 50ul models and below
Compatible with Finntip pipette tips, and other tip models
Specifications
Accuracy±3.0 to 0.6%
Compatible TipsFinntip 50 Micro, Filter 50 Micro
DescriptionFinnpipette F3 5-50μL micro
ColorTurquoise
ModelF3
Number of Channels1
Product LineFinnpipette™ F3
TypeVariable Volume Pipette
Volume (Metric)5 to 50 μL
Increments0.1μL
Precision2.5 to 0.3%
Color CodeTurquoise
Unit SizeEA