S58-A Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
S58-A Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
Actual product may vary
S58-A Fixed Angle Rotor for MTX and MX Plus Micro-Ultracentrifuges
Utilize the high gravitation force and 58,000rpm speed of the Thermo Scientific™ S58-A Fixed Angle Rotor.
Have Questions?
Catalog NumberCapacity
458668 x 13.5 mL
Catalog number 45866
Price (MYR)
97,971.00
EA
Add to cart
Capacity:
8 x 13.5 mL
Price (MYR)
97,971.00
EA
Add to cart
Utilize the high gravitation force and 58,000rpm speed of the Thermo Scientific™ S58-A Fixed Angle Rotor.
Compatible with:
Thermo Scientific™ Sorvall™ MTX 150 Series Micro-Ultracentrifuge, MX Series Floor Model Micro-Ultracentrifuge
Recommended for:
Subcellular isolations and pelleting.
Specifications
DescriptionS58-A Fixed Angle Rotor
MaterialAluminum
TypeFixed Angle Rotor
Capacity8 x 13.5 mL
Max. RCF288,959 x g
Max. Speed58,000 rpm
Radius (Metric)76.9 mm
Radius (Metric) Min.39.6 mm
K Factor50
Max. Tube Length18 mm
Max. Tube Diameter16 mm
Net Weight4.2 lb. (1.9 kg)
Angle35°
Avg. RCF219,067 x g
Min. RCF148,800 x g
BiocontainmentYes
Unit SizeEA