LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
Actual product may vary
LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
LinerGOLD™ GC Liners
Use the gold-standard Thermo Scientific™ LinerGOLD™ GC Liners for consistently high accuracy, sensitivity, and precision in your gas chromatography (GC) analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDiameter (Metric) InnerLength (Metric)Quantity
453T2120PTV Baffle Liner (Siltek)2 mm120 mm5
45322044PTV Siltek Metal Liner2 mm120 mm2
45350030Split Straight Liner5 mm105 mm5
45352057PTV Straight Liner2 mm120 mm5
45352062PTV Liner with Three Baffles1 mm120 mm5
453T1905Split FocusLiner for 50 mm Needle5 mm105 mm5
HYP453T2120PTV Baffle Liner (Siltek)2 mm120 mm5
45322045
also known as 45322045XXX
PTV Glass Liner2 mm120 mm5
453A1335-UILinerGOLD Direct Straight Liner1 mm78.5 mm5
HYP453A1335-UILinerGOLD Direct Straight Liner1 mm78.5 mm5
Catalog number 453T2120
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
PTV Baffle Liner (Siltek)
Diameter (Metric) Inner:
2 mm
Length (Metric):
120 mm
Quantity:
5
Use the gold standard in GC liner performance and consistency. LinerGOLD GC Liners provide enhanced inertness, leading to increased accuracy, sensitivity, and precision in your gas chromatography (GC) analysis. These injection port liners deliver:
  • Increased accuracy and sensitivity in analysis, preventing the loss of sensitive compounds in analysis
  • Lower detection limits when analyzing active and sensitive compounds
  • Excellent reproducibility gives confidence in your results from liner to liner
  • Consistently high level of performance time after time
LinerGOLD GC liners are manufactured and tested to ensure that you have confidence with every liner that is installed onto your GC system.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionPTV Baffle Liner (Siltek)
Quantity5
Length (Metric)120 mm
Packing MaterialNone
Product TypePTV Baffle Liner (Siltek)
Diameter (Metric) Inner2 mm
Diameter (Metric) Outer2.75 mm
For Use With (Equipment)Thermo Scientific Instruments
Unit SizeEach