Use these standard deactivated injection port liners to optimize performance of Thermo Scientific and Agilent GC instruments.
Have Questions?
Catalog Number
Diameter (Metric) Inner
Packing Material
Product Type
Quantity
453A2265
4 mm
Quartz Wool
Split Straight Liner
5
Catalog number 453A2265
HSN 73269099
Price (MYR)
980.00
PK
Add to cart
Diameter (Metric) Inner:
4 mm
Packing Material:
Quartz Wool
Product Type:
Split Straight Liner
Quantity:
5
Price (MYR)
980.00
PK
Add to cart
Optimize the performance of Thermo Scientific™ and Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
Highly deactivated borosilicate glass
Quartz wool packing material
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Application)Split
Quantity5
Length (Metric)78.5 mm
Packing MaterialQuartz Wool
Product TypeSplit Straight Liner
Diameter (Metric) Inner4 mm
Diameter (Metric) Outer6.3 mm
For Use With (Equipment)Thermo Scientific TRACE 1300/1600 series and Agilent GC instruments