Check Order Status
Dermacea™ 100% Cotton Gauze Sponges
Actual product may vary
Dermacea™ 100% Cotton Gauze Sponges
An alternative for sterile and nonsterile 100% cotton gauze dressings
Have Questions?
Catalog Number
Dimensions (L x W)
Thickness
Sterility
4412-11
2 x 2 in.
8-Ply
Sterile
Catalog number
4412-11
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Dimensions (L x W):
2 x 2 in.
Thickness:
8-Ply
Sterility:
Sterile
Specifications
Material
Cotton
Product Type
Gauze Sponge
Sterility
Sterile
Quantity
3000 Cs.
Dimensions (L x W)
2 x 2 in.
For Use With (Application)
Wound dressing, general cleaning, minor prepping, wound packing and debriding
Thickness
8-Ply