Bacterial Air Vents
Bacterial Air Vents
Actual product may vary
Bacterial Air Vents
For use as vents for receiving vessels or as sterile breathing barriers.
Have Questions?
Catalog NumberSterility
4308
also known as 4308JP
Non-sterile
Catalog number 4308
also known as 4308JP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Sterility:
Non-sterile
  • Economical and convenient depth filter for venting applications
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Material (Membrane)Glass Laminate
Pore Size1 μm
SterilityNon-sterile
Filtration Area7.5 cm2
Inlet StyleStepped Hose Barb
Outlet StyleStepped Hose Barb
Quantity10/Pk.
Diameter (Metric)37 mm
Unit SizeEA