Benchtop Dewar Flasks
Benchtop Dewar Flasks
Actual product may vary
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Benchtop Dewar Flasks
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with these benchtop Dewar flasks with double-walled sides, insulation and pour spouts.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
4150-900010 L
4150-20002 L
Catalog number 4150-9000
Price (MYR)
3,025.00
CS
Add to cart
Capacity (Metric):
10 L
Price (MYR)
3,025.00
CS
Add to cart
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.
  • High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
  • Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
  • Can be used from -196°C to +100°C
  • Pour spout and bottom grip area for ease of removal of used liquids
  • Vented, insulated cover allows for vapor escape but maintains sample temperature
Specifications
No. per Case1
No. per Pack1
Capacity (English)338.14 fl oz.
Capacity (Metric)10 L
Depth (Metric) Interior396 mm
Depth (English) Interior15.6 in.
Diameter (Metric) Inner Mouth198mm
Diameter (English) Inner Mouth7.8 in.
Height (Metric)470mm
Height (English)18.5 in.
Unit SizeCS