Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Actual product may vary
Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom: Sterile
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeClosure Type
4115-20002000 mL45-430 mmVented
FKS10592033250 mL38-430 mmVented
Catalog number 4115-2000
Price (MYR)
459.00
CS
Add to cart
Capacity (Metric):
2000 mL
Closure Size:
45-430 mm
Closure Type:
Vented
Price (MYR)
459.00
CS
Add to cart
Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features:
  • Sterile and single use: Reduce contamination risk
  • BPA-free
  • Collapses when autoclaved: Reduce bio hazardous waste
  • Graduated for quick volume assessment
  • Regulatory documentation available on request
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Capacity (English)64 oz.
Capacity (Metric)2000 mL
MaterialPETG
No. per Case4
No. per Pack1
ShapeErlenmeyer
Surface CoatingNone
AutoclavableNot Autoclavable
DisposableSingle-use
GraduatedYes
Mouth StyleNarrow
SterilitySterile
Bottom ShapePlain
Closure TypeVented
Closure Size45-430 mm
Unit SizeCS