Nalgene™ Polycarbonate Erlenmeyer Flasks
Nalgene™ Polycarbonate Erlenmeyer Flasks
Actual product may vary
Nalgene™ Polycarbonate Erlenmeyer Flasks
Nalgene™ Polycarbonate Erlenmeyer Flasks
Nalgene™ Polycarbonate Erlenmeyer Flasks
Nalgene™ Polycarbonate Erlenmeyer Flasks
Conduct many culturing techniques and store culture media with these flasks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
4103-0250250 mLStopper No. 6
4103-0125125 mLStopper No. 5
Catalog number 4103-0250
Price (MYR)
1,029.00
CS
Add to cart
Capacity (Metric):
250 mL
Closure Size:
Stopper No. 6
Price (MYR)
1,029.00
CS
Add to cart
Conduct many culturing techniques and store culture media with Thermo Scientific™ Nalgene™ Polycarbonate Erlenmeyer Flasks. The transparent wall provides a clear view of flask contents. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
  • Molded-in graduations
  • 500mL size is superior combustion flask for microdetermination of fluorine
  • Mouth sized to accept standard sized stoppers
  • Autoclavable/Transparent/Graduated
Note:  Repeated autoclaving may result in some loss of mechanical strength.   Wash and rinse well before autoclaving.
Specifications
Capacity (English)8 oz.
DescriptionFlask Erlenmeyer Graduated PC 250mL
Capacity (Metric)250 mL
MaterialPolycarbonate
No. per Case12
No. per Pack6
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
Bottom ShapePlain
Closure SizeStopper No. 6
Unit SizeCS