Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Actual product may vary
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
4102-2000
also known as NLG4102-2000
2000 mLStopper No. 10
Catalog number 4102-2000
also known as NLG4102-2000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
2000 mL
Closure Size:
Stopper No. 10
Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.
  • Translucent wall makes liquid level easily visible
  • 2000mL size has molded-in graduations
  • Mouth sized to accept standard sized stoppers (stopper sold separately)
  • Repeatedly autoclavable for long life and rigorous use
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications
Capacity (English)64 oz.
DescriptionFlask Erlenmeyer PP 2000mL
Capacity (Metric)2000 mL
MaterialPPCO
No. per Case6
No. per Pack2
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
Mouth StyleNarrow
SterilityNon-sterile
Bottom ShapePlain
Closure SizeStopper No. 10
Unit SizeCS