Check Order Status
Evolution 300 and 600 Spectrophotometer Smart Sipper System
Evolution 300 and 600 Spectrophotometer Smart Sipper System
Easy-to-use sampling accessory featuring a reliable multi-roller peristaltic pump
Have Questions?
Catalog Number
Type
401322844561
Silicone Pump Tubing
Catalog number
401322844561
Price (MYR)
-
Type:
Silicone Pump Tubing
For use with Thermo Scientific™ Evolution™ 300 and 600 Spectrophotometers
Complete and automated integration into VISION and Local Control software methods
Automatic recognition by software, smart alignment and serial number reporting
Handles sample volumes from 200μL to 9.999mL with customizable sip volume and air gap
Methods can be automated for sample measurement after they are sipped
Specifications
Type
Silicone Pump Tubing
For Use With (Equipment)
Evolution™ 300 and 600 spectrophotometer Smart Sipper System