Search Thermo Fisher Scientific
| Catalog Number | Color |
|---|---|
| 4000222 | Black |
| 4000221 | Multi-color |
Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.