Cryogenic Box Dividers
Cryogenic Box Dividers
Actual product may vary
Cryogenic Box Dividers
Maximize storage capacity with Thermo Scientific™ Croygenic Boxes, for use in Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage.
Have Questions?
Catalog NumberFor Use With (Equipment)TypeHoldsVial Capacity
4000014CryoPlus 1,2,3,4Fiberboard Box2 mL Vials with 100-cell Divider100
Catalog number 4000014
Price (MYR)
60.00
EA
Add to cart
For Use With (Equipment):
CryoPlus 1,2,3,4
Type:
Fiberboard Box
Holds:
2 mL Vials with 100-cell Divider
Vial Capacity:
100
Price (MYR)
60.00
EA
Add to cart
Maximize storage capacity with Thermo Scientific™ Croygenic Box Dividers, for use in Thermo Scientific™ CryoPlus™ LN2 cryopreservation storage.
For Cardboard boxes
  • Vertical (2mL with 100 cell divider)
  • Square (2, 4 or 5mL with 100 cell divider)
  • Arrowhead (2, 4 or 5mL with 81 cell divider)
For Polycarbonate boxes
  • Vertical (2mL with 100 or 81 cell division)
  • Square (2mL with 100 or 81 cell division; 4mL with 81 cell division)
For Stainless-steel boxes†
  • Square (2 or 4mL)
  • Arrowhead (2mL)
  • †Stainless steel boxes do not include cell dividers
  • Warranty varies by country, please contact us for details
Specifications
TypeFiberboard Box
Capacity (English)7.06 cu. ft.
Capacity (Metric)2 mL
For Use With (Equipment)CryoPlus 1,2,3,4
Vial Capacity100
Holds2 mL Vials with 100-cell Divider
Unit SizeEA