Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Actual product may vary
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Cryo Vial Closure Color Coders
Make it easier to identify specific vials quickly and simplify your inventory by using these color coders that fit into the top of cryogenic vial or tube closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorProduct Line
375884PinkNunc
354755WhiteNunc
354879BlueNunc
354968
also known as NNC354968
RedNunc
355018GreenNunc
355077
also known as NNC355077
YellowNunc
375868
also known as NNC375868
BrownNunc
375906
also known as NNC375906
GrayNunc
375922
also known as NNC375922
PurpleNunc
5045-0003BlueNalgene
5045-0000
also known as 5045-0000PNS
WhiteNalgene
5045-0002
also known as 5045-0002PNS
YellowNalgene
5045-0004GreenNalgene
5045-0005RedNalgene
Catalog number 375884
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Pink
Product Line:
Nunc
Make it possible to identify specific vials quickly and simplify your inventory using color coders that fit into the top of Nalgene™ Cryogenic vial closures or Nunc™ Cryogenic Tube closures.
  • Nalgene™ color coders are made from polystyrene and fit only in Nalgene™ cryogenic vial closures. Available in 5 colors
  • Nunc™ color coders are made from polycarbonate and fit only in Nunc™ cryogenic tube closures and Cryobank 2ml and 5mL tube caps. Available in 10 colors or as an assortment.
Specifications
DescriptionNunc™ CryoColor Vial Closure Coder, PS
ColorPink
MaterialPolycarbonate
Product LineNunc
Quantity500/Pk., 2000/Cs.
TypeVial Closure
Unit SizeCS