Pre-cut PEEK Tubing in a 5-foot Coil
Pre-cut PEEK Tubing in a 5-foot Coil
Actual product may vary
Pre-cut PEEK Tubing in a 5-foot Coil
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) Inner
370400.040 in.
370070.007 in.
Catalog number 37040
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (English) Inner:
0.040 in.
Thermo Scientific™ Pre-cut PEEK Tubing in a 5-foot Coil is easily cut to the desired length.
  • Broad chemical compatibility
  • Biocompatible
  • May be used with standard fingertight fittings
  • Resistant to most organic solvents however nitric acid, sulfuric acid, dichloromethane, THF and DMSO are not recommended
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use WithHPLC
TypeHPLC Tubing
Description0.040 in. ID; 5 ft. length; Natural
For Use With (Application)HPLC
Diameter (English) Inner0.040 in.
Diameter (Metric) Inner1.016 mm
Overall Length (English)5 ft.
Overall Length (Metric)1.524 m
MaterialPEEK
Unit SizeEach