PA Thinwall Tube
PA Thinwall Tube
Actual product may vary
PA Thinwall Tube
PA Thinwall Tube for ultraspeed centrifuge.
Have Questions?
Catalog NumberFor Use With (Equipment)
3699
also known as QSF3699
Ultraspeed Centrifuge
Catalog number 3699
also known as QSF3699
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Ultraspeed Centrifuge
  • PA Thinwall Tube, 13.2 mL x 50, for ultraspeed centrifuge
Specifications
TypeThinwall Tube
MaterialNylon
For Use With (Equipment)Ultraspeed Centrifuge
DisposableReusable
Capacity50 x 13.2 mL
Unit SizeEA