Replacement Needles for GC Syringes
Replacement Needles for GC Syringes
Actual product may vary
Replacement Needles for GC Syringes
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Have Questions?
Catalog NumberFor Use With (Equipment)
365RN235Thermo 365K3041 Syringe
Catalog number 365RN235
Price (MYR)
-
For Use With (Equipment):
Thermo 365K3041 Syringe
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description365RN235
Needle Gauge23 G
Length (English) Needle1.9685 in.
Product TypeReplacement Syringe Needle
Volume (Metric)1 mL
Length (Metric) Needle50 mm
Needle Point StyleBevel
For Use With (Equipment)Thermo 365K3041 Syringe
Connection TypeRemovable
Unit SizePack