Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Actual product may vary
Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Pick the optimal choice for a busy lab with these dedicated platforms, which are model-specific and come with pre-installed clamps to maximize capacity.
Have Questions?
Catalog NumberProduct TypeHoldsDimensions (L x W)For Use With (Equipment)
3524-60Adjustable Vessel Platform18 x 18 in. (45.7 x 45.7 cm)MaxQ™ 4000 and 6000 Shaker
Catalog number 3524-60
Price (MYR)
12,114.00
EA
Add to cart
Product Type:
Adjustable Vessel Platform
Dimensions (L x W):
18 x 18 in. (45.7 x 45.7 cm)
For Use With (Equipment):
MaxQ™ 4000 and 6000 Shaker
Price (MYR)
12,114.00
EA
Add to cart
Pick the optimal choice for a busy lab with Thermo Scientific™ Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers. MaxQ platforms are model-specific and come with pre-installed clamps to maximize capacity. MaxQ shakers, platforms and accessories provide exceptional innovation, reliability and accuracy.
Clamps come installed to provide maximum capacity.
Specifications
Product TypeAdjustable Vessel Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)18 x 18 in. (45.7 x 45.7 cm)
Length (English) Platform18 in.
Length (Metric) Platform45.7 cm
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
Description6000 shakers
For Use With (Equipment)MaxQ™ 4000 and 6000 Shaker
Unit SizeEA