EK Immersion Coolers
EK Immersion Coolers
Actual product may vary
EK Immersion Coolers
EK Immersion Coolers
EK Immersion Coolers
Simplify direct cooling of small samples with immersion coolers. Combine immersion coolers with open-bath circulators for a reliable, flexible alternative to tap water cooling that eliminates dry ice or liquid nitrogen handling--saving both time and money.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCooling CapacityTemperature RangeVoltage
3291901350w at +20°C; 280w at -10°C; 170w at -40°C; 100w-90°C to +40°C230 V
329-1901350w at +20°C; 280w at -10°C; 170w at -40°C; 100w-90°C to +40°C230 V
Catalog number 3291901
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Cooling Capacity:
350w at +20°C; 280w at -10°C; 170w at -40°C; 100w
Temperature Range:
-90°C to +40°C
Voltage:
230 V
A simple solution for direct cooling of small samples. Thermo Scientific™ EK Immersion Coolers offer wide temperature ranges and integrated control. They can be used with open-bath circulators to provide the easiest way to cool small samples. A reliable and flexible alternative to tap water cooling that eliminates the inconvenience of handling dry ice or liquid nitrogen, saving both time and money. All models feature CFC-free refrigerant.
Models EK45 and EK90
  • Multi-purpose coolers with own controller for temperature accuracy of approximately 1° to 2°C
  • Suitable for individual cooling applications like cooling smaller volumes or removing reaction heat
  • Air-cooled cooling circuit Digital temperature display
  • EK45: Reaches temperatures down to -45°C in a 5L Dewar vessel
  • EK90: Reaches temperatures down to -90°C in a 5L Dewar vessel
  • Warranty varies by country, please contact us for details
Specifications
DescriptionEK 90 Temperature Setpoint Option, Flexible Probe, 230V/50Hz
Cooling Capacity350w at +20°C; 280w at -10°C; 170w at -40°C; 100w
Temperature Range-90°C to +40°C
ModelEK90
Length (English) Hose59 in.
Length (Metric) Hose150 cm
Electrical Requirements230 V 50/60 Hz