Forma™ Series II Water-Jacketed CO2 Incubator, 184L
Forma&trade; Series II Water-Jacketed CO<sub>2</sub> Incubator, 184L
Actual product may vary
Forma&trade; Series II Water-Jacketed CO<sub>2</sub> Incubator, 184L
Forma&trade; Series II Water-Jacketed CO<sub>2</sub> Incubator, 184L
Forma&trade; Series II Water-Jacketed CO<sub>2</sub> Incubator, 184L
Forma™ Series II Water-Jacketed CO2 Incubator, 184L
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series II 3110 Water-Jacketed CO2 Incubators.
Have Questions?
Catalog NumberdimVoltageCO2 Sensor TechnologyOxygen Control
3141IR Sensor1 to 21%
Catalog number 3141
Price (MYR)
-
CO2 Sensor Technology:
IR Sensor
Oxygen Control:
1 to 21%
Thermo Scientific™ Forma™ Series II Water-Jacketed CO2 Incubators provide precise CO2 control, unsurpassed temperature stability and superior parameter recovery characteristics, with continuous contamination control technology. They provide an environment for the growth of microbiological samples or mammalian cells for research (FDA Class 1), and IVF and other regulated cell therapy applications (FDA Class 2).
Water Jacketed Chamber
  • The 184L (6.5 cu. ft.) high capacity chamber provides outstanding culturing space
  • Standard right hand door swing, field reversible 
  • Polished stainless steel interior with coved corners is easy-to-clean with removable humidity pan
  • Unique triple wall construction provides outstanding temperature uniformity and stability
  • Exclusive, heated dual pane inner glass door is designed to minimize condensation
  • In-chamber fan gently distributes air throughout the chamber which is designed ensure homogeneous conditions
  • Sturdy stainless steel shelves and supports can be removed without tools for easy cleaning or adjustment
ISO Class 5 HEPA Filtration Airflow System
  • In-chamber HEPA air filtration continuously filters entire chamber volume every 60 seconds
  • Reaches ISO Class 5 (Class 100) cleanroom air quality in only 5 minutes after door opening
  • Continuous protection by minimizing ability for contaminants to settle on interior surfaces
In situ Sensors and Probes Designed for Precise Monitoring
  • Choice of  TC (thermal conductivity) CO2 sensor for long, reliable operation or  IR (infrared) CO2 sensor  for monitoring when humidity and temperature are less predictable
  • Dual temperature probes with over temperature protection
  • Optional rH display for monitoring humidity levels to prevent “dry-outs"
Enviro-Scan Microprocessor Control
  • Easy- to-use, alphanumeric LED display
  • Flexible, intuitive controls with message center including process and alarm status messages
  • Configurable audible on/off, access code, tracking low and high alarms
Specifications
Capacity (English)6.5 cu. ft.
Certifications/ComplianceUL, cUL, CE
Capacity (Metric)184 L, 184 L
For Use WithFor use with T-865, T-1250, Superspin 360 (36 mL) and AH-629 (36 mL) rotors
Data OutputsOptional 4-20mA
No. per Pack25
DescriptionSingle Tri-gas 184L incubator, 1-21%
Chamber MaterialPolished Stainless Steel
CO2 Concentration Range0 to 20%
CO2 Sensor TechnologyIR Sensor
LightingLED
MaterialStainless Steel
Oxygen Control1 to 21%
Relative Humidity<90% at 37°C
Temperature Range (Metric)Ambient +5° to 50°C
TypeCO2 Incubator
Product TypeWalled Tube
Max. RCF429,459 x g
Humidity SourceWater Pan
Dimensions (D x W x H) Interior20 x 21.3 x 26.8 in. (50.8 x 54.1 x 68.1 cm)
Weight (English)365 lb.
Electrical Requirements230 V, 50/60 Hz
Unit SizeEA