Nalgene™ Wide-Mouth HDPE Centrifuge Bottles
Nalgene™ Wide-Mouth HDPE Centrifuge Bottles
Actual product may vary
Nalgene™ Wide-Mouth HDPE Centrifuge Bottles
Carry out applications such as large-volume cell harvesting, pelleting and protein purification with Thermo ScientificTM NalgeneTM Wide-Mouth HDPE Centrifuge Bottles.
Have Questions?
Catalog NumberNominal CapacityMax. Force
3121-0250
also known as 3121-0250JP
250 mL8000 x g
Catalog number 3121-0250
also known as 3121-0250JP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Nominal Capacity:
250 mL
Max. Force:
8000 x g
Carry out applications such as large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Centrifuge Bottles, molded of rigid and robust high-density polyethylene. These translucent bottles have excellent chemical resistance properties and are rated for temperatures as low as -100°C.
Features:
  • For low speed centrifugation of biological and chemical samples
  • Excellent for use in refrigerated centrifuges
  • Rated for forces up to 8000 x g
  • For proper performance bottles, must be filled to at least 80% of total capacity
Requires:
When spinning 250mL bottle in a Sorvall™ GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required
Specifications
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorTranslucent
DescriptionHigh Density Polyethylene Bottle with PP Closure
MaterialHDPE
Length (Metric)131.6 mm
No. per Case36
No. per Pack6
Nominal Capacity250 mL
GraduatedNo
Max. Force8000 x g
MouthWide
ShapeRound
Height (Metric)131.6 mm
Product TypeCentrifuge Bottle
Closure MaterialPolypropylene
Diameter (Metric)61.2 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeCS