Hypersil™ APS-2 HPLC Columns, 5μm Particle Size
Hypersil™ APS-2 HPLC Columns, 5μm Particle Size
Actual product may vary
Hypersil™ APS-2 HPLC Columns, 5μm Particle Size
Analyze sugars with this versatile amino propyl phase of Thermo ScientificTM HypersilTM APS-2 HPLC Columns.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
30705-254030
also known as 30705-254030B
250 mm4 mm5 μm
Catalog number 30705-254030
also known as 30705-254030B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric):
4 mm
Particle Size:
5 μm
Analyze sugars with this versatile amino propyl phase of Thermo ScientificTM HypersilTM APS-2 HPLC Columns.
Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM Amino columns as an alternative.
  • In reversed phase mode, Hypersil APS-2 columns are excellent for carbohydrate analysis: extra sensitivity for sugars
  • Alternative selectivity to silica when used in normal phase
  • Ordering Information:
    Other column dimensions are available. Also available in 10μm. Please call your Customer Service Representative for more information.
    Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Column FormatAnalytical Column
    Diameter (Metric)4 mm
    For Use WithIC
    Length (Metric)250 mm
    Particle Size5 μm
    pH2.5 to 8
    Pore Size120 Å
    Surface Area170 m²/g
    Temperature60°C
    USP TypeL8
    Stationary PhaseAmino (NH2)
    Product LineHypersil Classical
    Packing MaterialSpherical, Fully Porous Type A Silica
    Carbon Load1.9%
    Column TypeCarbohydrate Analysis
    EndcappedNo
    PhaseReversed Phase, Normal Phase, Ion Exchange, HILIC
    Unit SizeEach