Test Tube Racks for MaxQ™ Shaker
Test Tube Racks for MaxQ™ Shaker
Actual product may vary
Test Tube Racks for MaxQ™ Shaker
Mix and match these shaker test tube racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHoldsArray
30189GreenTubes (26 to 30 mm), Centrifuge Tubes (50 mL)3 x 3
30180BIRedTubes (10 to 13 mm)6 x 12
30182OrangeTubes (14 to 16 mm), Centrifuge Tubes (15 mL)6 x 12
30185WhiteTubes (17 to 20 mm)4 x 5
30184WhiteTubes (17 to 20 mm)4 x 10
30188GreenTubes (26 to 30 mm)3 x 8 array
Catalog number 30189
Price (MYR)
3,171.00
EA
Add to cart
Color:
Green
Holds:
Tubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Array:
3 x 3
Price (MYR)
3,171.00
EA
Add to cart
Mix and match Thermo Scientific™ MaxQ™ Shaker Test Tube Racks, available in a variety of sizes and arrays, to suit individual needs in the laboratory. All racks are compatible with MaxQ shaker platforms.
Racks are available in a multitude of sizes, arrays, and colors. Compatible with MaxQ shaker platforms.
Specifications
Product TypeTest Tube Rack
DescriptionGreen; 26 to 30 mm; 3 x 3 array (fits 50 mL centrifuge tubes)
Array3 x 3
ColorGreen
HoldsTubes (26 to 30 mm), Centrifuge Tubes (50 mL)
Positions9
For Use With (Equipment)MaxQ™ Shaker Platform
Unit SizeEA