Check Order Status
500 μg/mL Acrylamide-d3 in Acetonitrile, Restek
Actual product may vary
500 μg/mL Acrylamide-d3 in Acetonitrile, Restek
Have Questions?
Catalog Number
Quantity
30153
5 mL
Catalog number
30153
Price (MYR)
348.00
EA
Add to cart
Quantity:
5 mL
Price (MYR)
348.00
EA
Add to cart
Deuterium-labeled acrylamide
Cost-effective isotopically labeled internal standard choice for analysis of acrylamide in food or environmental samples
Specifications
Quantity
5 mL
Concentration
500 μg/mL
Certifications/Compliance
Certified reference material
Solvent or Matrix
Acetonitrile
CAS
122775-19-3