Check Order Status
2-Furoyl-LIGRLO-amide
Actual product may vary
2-Furoyl-LIGRLO-amide
Potent and selective PAR2 agonist
Have Questions?
Catalog Number
Quantity
30151
1 mg
Catalog number
30151
Price (MYR)
303.00
EA
Add to cart
Quantity:
1 mg
Price (MYR)
303.00
EA
Add to cart
2-Furoyl-LIGRLO-amide is a potent and selective PAR2 receptor agonist (pD2 = 7.0). Causes a dose-dependent relaxation of murine femoral arteries.
Specifications
Purity
0.95
Quantity
1 mg