Glow-In-The-Dark/Ultra Reflective Number: 0
Glow-In-The-Dark/Ultra Reflective Number: 0
Actual product may vary
Glow-In-The-Dark/Ultra Reflective Number: 0
Identify equipment, bins and areas of warehouse required by facility safety and maintenance. Brady™ Glow-In-The-Dark/Ultra Reflective Number signs reflect light and glow brightly, combining a high level of reflectivity with advanced glow-in-the-dark technology.
Have Questions?
Catalog NumberSize (English)
30100
also known as EME30100/CEB
2.5 x 3.5 in. (W x H)
Catalog number 30100
also known as EME30100/CEB
Price (MYR)
2,028.00
EA
Add to cart
Size (English):
2.5 x 3.5 in. (W x H)
Price (MYR)
2,028.00
EA
Add to cart
  • Micro-prism reflectors for maximum visibility from greater distance, wider angle and lower light source
  • Designed for permanent Indoor/Outdoor applications
  • Made of high intensity prismatic reflective sheeting photoluminescent material with pressure sensitive adhesive
Specifications
Adhesive TypeAcrylic, Permanent
ColorBlack, Silver, Green, Glow
MaterialVinyl (B-309)
Product TypeUltra Reflective Number Label
Display TextLegend: 0
For Use With (Application)Labeling in Low Light Conditions (Indoor/Outdoor)
Size (English)2.5 x 3.5 in. (W x H)
Size (Metric)6.3 x 8.89cm
Height (English) Character3 in.
Unit SizeEA