Ferrules for Agilent™ Instruments
Ferrules for Agilent™ Instruments
Actual product may vary
Ferrules for Agilent™ Instruments
Ensure reliable connections for Agilent™ GC instruments with these Thermo Scientific™ Ferrules for Agilent Instruments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialDiameter (Metric)
290VA19315% Graphite, 85% Vespel0.45 to 0.53 mm
290GA139
also known as 072635TH
100% Graphite0.1 to 0.32 mm
290GA140100% Graphite0.45 to 0.53 mm
290VA19115% Graphite, 85% Vespel0.1 to 0.25 mm
290VA19215% Graphite, 85% Vespel0.32 mm
HYP290VA19215% Graphite, 85% Vespel0.32 mm
Catalog number 290VA193
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
15% Graphite, 85% Vespel
Diameter (Metric):
0.45 to 0.53 mm
Use Thermo Scientific™ Ferrules for Agilent™ Instruments to ensure optimal performance of your Agilent GC instrument. They are available in two different materials of various dimension to accommodate a range of instruments and applications.
  • 100% Graphite ferrules
  • 15% graphite/85% vespel
Ordering Information
Packaged in contaminant-free, individual blister packs.
Specifications
Diameter (Metric)0.45 to 0.53 mm
For Use With (Equipment)Agilent Injectors and Non-MS Detectors
Material15% Graphite, 85% Vespel
TypeFerrule
QuantityPack of 10
Unit SizePack