Finger Tite Ferrules
Finger Tite Ferrules
Actual product may vary
Finger Tite Ferrules
Finger Tite Ferrules
Finger Tite Ferrules
Finger Tite Ferrules
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Have Questions?
Catalog NumberDiameter (Metric)
290S11320.4 mm
Catalog number 290S1132
Price (MYR)
-
Diameter (Metric):
0.4 mm
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific Finger Tite ferrules are the next generation of the SilTite™ connection system.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • No tools required
Specifications
Description0.4 mm ferrule
Diameter (Metric)0.4 mm
For Use With (Equipment)0.1 to 0.25 mm I.D. GC Columns
TypeFerrule
Unit SizePack