GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Actual product may vary
GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Have Questions?
Catalog NumberDescriptionFor Use With (Equipment)
290OA241
also known as 0726532TH
Viton Liner Sealing RingTRACE 1300 Series GC and Agilent SSL Injectors
Catalog number 290OA241
also known as 0726532TH
Price (MYR)
-
Description:
Viton Liner Sealing Ring
For Use With (Equipment):
TRACE 1300 Series GC and Agilent SSL Injectors
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
Specifications
DescriptionViton Liner Sealing Ring
Quantity10 Pack
Product TypeViton Liner Sealing Ring
For Use With (Equipment)TRACE 1300 Series GC and Agilent SSL Injectors
Unit SizePack