GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Actual product may vary
GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDiameter (Metric) InnerFor Use With (Equipment)Quantity
290GA082Gold Inlet Base Seal0.8 mmSingle Column Installation2 Pack
290GA081Gold Inlet Base Seal0.8 mmSingle Column Installation10 Pack
Catalog number 290GA082
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Gold Inlet Base Seal
Diameter (Metric) Inner:
0.8 mm
For Use With (Equipment):
Single Column Installation
Quantity:
2 Pack
Ensure optimal gas chromatography (GC) analysis by using exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals. Precision machined from high grade stainless steel, choose from gold-plated or Siltek-treated base seals. The chemically inert coated surface makes them suitable for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenols. Use these GC Injection Port Base Seals with split/splitless injection ports of thermo Scientific™ TRACE™ 1300 Series GC and Agilent GC 5890/6890/7890 systems.
  • Precision machined to provide exceptional sealing properties
  • Reduces activity (especially useful for analysis of pesticide, PCBs, phenols, etc.)
  • High grade stainless steel provides reproducible seal
  • Direct equivalent to Agilent original part
  • Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionGold Inlet Base Seal
Quantity2 Pack
Product TypeGold Inlet Base Seal
Diameter (Metric) Inner0.8 mm
For Use With (Equipment)Single Column Installation
Unit SizePack