Hypersil™ BDS C8 HPLC Columns
Hypersil™ BDS C8 HPLC Columns
Actual product may vary
Hypersil™ BDS C8 HPLC Columns
Use as a robust, general-purpose column for QC/QA applications. They are high-quality, base-deactivated, fully endcapped, have similar C18 selectivity but slightly less retentive.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle SizeUnit Size
28205-124630125 mm4.6 mm5 μmEach
28205-154630150 mm4.6 mm5 μmEach
28205-254030250 mm4 mm5 μmEach
28205-254630
also known as HYP28205-254630
250 mm4.6 mm5 μmEach
HYP28205-254630250 mm4.6 mm5 μmEA
28205-01400110 mm4 mm5 μmEach
28203-154630
also known as HYP28203-154630
150 mm4.6 mm3 μmEach
HYP28203-154630150 mm4.6 mm3 μmEA
Catalog number 28205-124630
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
125 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Unit Size:
Each
For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.
  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
For Use WithHPLC
Length (Metric)125 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 9
Pore Size130 Å
Surface Area170 m²/g
Temperature60°C
USP TypeL7
Stationary PhaseC8
Product LineHypersil BDS
Packing MaterialSpherical, Fully Porous, Base-Deactivated Silica
Carbon Load7%
Column TypeReversed Phase
EndcappedYes
PhaseReversed Phase
Unit SizeEach