Bead Ruptor 96 50 mL Milling Jars and O-rings Set of 2
Bead Ruptor 96 50 mL Milling Jars and O-rings Set of 2
Actual product may vary
Bead Ruptor 96 50 mL Milling Jars and O-rings Set of 2
Bead Ruptor 96 Milling Jars and O-rings are designed for rapid and efficient disruption of larger samples.
Have Questions?
Catalog number 27006
Price (MYR)
-
  • 2 stainless steel milling jars and o-rings
  • The jars are sealed via a screw mechanism with an o-ring interface.
  • Stainless steel jars are ideal for dry grinding or cryomilling applications of very tough material including, bone, seeds, paper and plastics.
Specifications
Product TypeMilling Jars
AutoclavableAutoclavable
MaterialStainless Steel
For Use With (Application)Ideal for dry grinding or cryomilling applications.
For Use With (Equipment)Bead Ruptor 96
Holds8 to 20 mL
Sample TypeVery tough material including, bone, seeds, paper and plastics
Unit SizeEach