Evolution™ Smart™ Sipper Accessories
Evolution™ Smart™ Sipper Accessories
Actual product may vary
Evolution™ Smart™ Sipper Accessories
Evolution™ Smart™ Sipper Accessories
Evolution™ Smart™ Sipper Accessories
Get the most out of your Thermo Scientific™ Evolution™ Spectrophotometer with a wide selection of Thermo Scientific™ Evolution™ Smart™ Sipper Accessories.
Have Questions?
Catalog NumberType
268-857400Quartz Flowcell, 10 mm path
Catalog number 268-857400
Price (MYR)
7,572.00
EA
Add to cart
Type:
Quartz Flowcell, 10 mm path
Price (MYR)
7,572.00
EA
Add to cart
Thermo Scientific™ Evolution™ Smart™ Sipper Accessories for Thermo Scientific™ Evolution™ Spectrophotometers are easy-to-use sampling accessories featuring a reliable multi-roller peristaltic pump. When the Smart Sipper is installed, all sipper controls are automatically displayed in the software. Methods can be automated so that samples are measured automatically after they are sipped. Smart Sipper Accessories feature modes to return the sample to the original container, send the sample to waste, or run continuously.
  • Automatic recognition by software, smart alignment and serial number reporting.
  • Able to handle sample volumes from 200μL to 9.999mL and a fully customizable sip volume and air gap.
  • Software assisted calibration routine to simplify the process of changing tubing and methods.
  • Sippers support 10mm pathlength cells and shorter pathlength cells in the standard cell holder with use of an appropriate spacer. Swap in a long path cell holder to support flow cells up to 100mm pathlength.
  • Specialized tubing packs available for:
    • mild chemical use (silicone tubing)
    • acid resistant (c-flex)
    • bio-compatible (Chemdurance Bio)
    • chemical resistant (Viton)
  • Replacement flow-cell connector packs for both Starna and Hellma cells available.
Ordering Alerts:
When deploying the Smart Sipper Accessory in a regulated or quality control environment, take advantage of our unique full-system qualification to assure compliance of the instrument and accessory as a complete unit.
Specifications
TypeQuartz Flowcell, 10 mm path
For Use With (Equipment)Evolution Smart Sippers
IncludesScrew connectors and PTFE tubing
Unit SizeEA