Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Actual product may vary
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
Have Questions?
Catalog NumberColorSterilityWell ShapeFit for Purpose
267245NaturalNon-sterileRoundCertified Quality
Catalog number 267245
Price (MYR)
1,835.00
CS
Add to cart
Color:
Natural
Sterility:
Non-sterile
Well Shape:
Round
Fit for Purpose:
Certified Quality
Price (MYR)
1,835.00
CS
Add to cart
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
  • Shared wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase and DNase Free. ANSI Compliant.
FormatStandard
LidWithout Lid
Description96-well Polypropylene Sample Processing & Storage Plates
Array8 x 12
AutoclavableAutoclavable
Packaging10 pk., 120 cs.
Binding TypeLow binding PP
Product LineNunc
ColorNatural
Product Type96-well plate
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
MaterialPolypropylene
Volume (Metric) Well500 μL
No. of Wells96
Volume (Metric) Working450 μL
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeRound
Dnase FreeYes
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
No. per Case120
No. per Pack10
Quantity120/Cs.
Regulatory StatusFor Research Use Only. Not for use in diagnostic procedures.
RNase FreeYes
Fit for PurposeCertified Quality
Plate SizeANSI/ SLAS Standard
Unit SizeCS