TraceGOLD TG-VRX GC Columns
TraceGOLD TG-VRX GC Columns
Actual product may vary
TraceGOLD TG-VRX GC Columns
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
26081-332030 m0.25 mm
Catalog number 26081-3320
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
30 m
Diameter (Metric):
0.25 mm
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
  • Proprietary stationary phase
  • Optimized dimensions for analysis of volatile organic pollutants
  • Ideally suited for US EPA Methods 502.2, 524.2, 8021, and 8260
  • Similar to DB-VRX, Rtx-VRX
Recommended for:
  • Volatile organic pollutants
  • Aromatics
Specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-VRX
Max. Temperature260°C
For Use With (Application)Volatile Organic Pollutants Analysis, GC/MS for US EPA Method 8021 Compounds
Film Thickness1.4 μm
Length (Metric)30 m
PolarityNon-Polar
Description30 m x 0.25 mm x 1.40 μm
Unit SizeEach