TraceGOLD™ TG-SQC GC Columns
TraceGOLD™ TG-SQC GC Columns
Actual product may vary
TraceGOLD™ TG-SQC GC Columns
Ensure the quality of your Thermo Scientific instrument when installed on site.
Have Questions?
Catalog NumberLength (Metric)
26070-130015 m
Catalog number 26070-1300
Price (MYR)
-
Length (Metric):
15 m
Ensure the quality of your Thermo Scientific instrument when installed on site by using the Thermo Scientific™ TraceGOLD™ TG-SQC GC column. The TraceGOLD TG-SQC GC column is optimized for system qualification tests during initial installation, or during service and maintenance of an installed instrument. This column is recommended for benchmark testing only.
Applications:
System qualification tests (Thermo Scientific GC-MS)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Length (Metric)15 m
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD TG-SQC
Functionality/Interaction TypeBenchmark Testing Only
Product LineTraceGOLD
Film Thickness0.25 μm
QuantityEach
Unit SizeEach