Hypersil GOLD™ Cyano HPLC Columns
Hypersil GOLD™ Cyano HPLC Columns
Actual product may vary
Hypersil GOLD™ Cyano HPLC Columns
Alternate selectivity with lower hydrophobicity. Use these HPLC columns for normal and reversed phase separations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
25805-254630
also known as 25805-254630RP
250 mm4.6 mm5 μm
25805-154630
also known as 25805-154630B
150 mm4.6 mm5 μm
Catalog number 25805-254630
also known as 25805-254630RP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
250 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Alternate selectivity with lower hydrophobicity. Use Thermo Scientific™ Hypersil Gold™ Cyano LC Columns for normal and reversed phase separations.
  • Excellent peak shape
  • Outstanding sensitivity
  • Less retention for faster analysis
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
USP TypeL10
Stationary PhaseCyano (CN)
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load4%
Column TypeMixed Mode
EndcappedYes
PhaseNormal and Reversed Phase
TypeAnalytical Column
Unit SizeEach