Alternate selectivity with lower hydrophobicity. Use these HPLC columns for normal and reversed phase separations.
Have Questions?
Change view
Catalog Number
Length (Metric)
Diameter (Metric)
Particle Size
25805-154630
also known as 25805-154630B
150 mm
4.6 mm
5 μm
25805-254630
also known as 25805-254630RP
250 mm
4.6 mm
5 μm
2 Options
Catalog number 25805-154630
also known as 25805-154630B
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Particle Size:
5 μm
Alternate selectivity with lower hydrophobicity. Use Thermo Scientific™ Hypersil Gold™ Cyano LC Columns for normal and reversed phase separations.
Excellent peak shape
Outstanding sensitivity
Less retention for faster analysis
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.