MaxQ™ HP Orbital Shakers Universal Platforms
MaxQ™ HP Orbital Shakers Universal Platforms
Actual product may vary
MaxQ™ HP Orbital Shakers Universal Platforms
Mix and match different size labware on a single platform with these orbital shakers universal platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDimensions (L x W)For Use With (Equipment)
238061Universal Platform without Clamps, 18 x 29.5 in.18 x 29.5 in. (74.9 x 45.7 cm)MaxQ 8000
238000Universal Platform without Clamps, 18 x 18 in.18 x 18 in.MaxQ™ HP Orbital Shakers
Catalog number 238061
Price (MYR)
22,038.00
EA
Add to cart
Description:
Universal Platform without Clamps, 18 x 29.5 in.
Dimensions (L x W):
18 x 29.5 in. (74.9 x 45.7 cm)
For Use With (Equipment):
MaxQ 8000
Price (MYR)
22,038.00
EA
Add to cart
Mix and match different size labware on a single platform with flexible Thermo Scientific™ MaxQ™ HP Orbital Shakers Universal Platforms.
Specifications
Product TypeShaker Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)18 x 29.5 in. (74.9 x 45.7 cm)
Length (English) Platform29.5 in.
Length (Metric) Platform74.9 cm
Width (English) Platform18 in.
Width (Metric) Platform45.7 cm
DescriptionUniversal Platform without Clamps, 18 x 29.5 in.
For Use With (Equipment)MaxQ 8000
Unit SizeEA