7961 Series Thermal Transfer Printer Ribbon
7961 Series Thermal Transfer Printer Ribbon
Actual product may vary
7961 Series Thermal Transfer Printer Ribbon
Thermal transfer ribbon with excellent scratch and good chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)Diameter (Metric)
236016300 m170 mm25.4 mm
236013300 m60 mm25.4 mm
236014300 m110 mm25.4 mm
236015300 m114 mm25.4 mm
Catalog number 236016
Price (MYR)
505.00
EA
Add to cart
Length (Metric):
300 m
Width (Metric):
170 mm
Diameter (Metric):
25.4 mm
Price (MYR)
505.00
EA
Add to cart
When printed on the appropriate Brady™ label, this ribbon gives excellent scratch and good chemical resistance.
Specifications
ColorBlack
For Use With (Equipment)BradyPrinter PR Plus Series Label Printer, THT 76 mm Core Printer
MaterialResin
Length (English)984 ft.
Width (English)6.7 in.
Length (Metric)300 m
Width (Metric)170 mm
Diameter (English)1 in.
Diameter (Metric)25.4 mm
FormatOpen-core Ribbon
Certifications/Compliance2011/65/EU and 2015/863
Alternate Part NumberR7961-170X300/O
Unit SizeEA