HyPURITY™ C18 HPLC Columns, 3μm Particle Size
HyPURITY™ C18 HPLC Columns, 3μm Particle Size
Actual product may vary
HyPURITY™ C18 HPLC Columns, 3μm Particle Size
Thermo ScientificTM HyPURITYTM C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
22103-154030150 mm4 mm3 μm
22103-153030150 mm3 mm3 μm
22105-154630
also known as 22105-154630B
150 mm4.6 mm5 μm
22105-254630
also known as 22105-254630B
250 mm4.6 mm5 μm
Catalog number 22103-154030
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
Diameter (Metric):
4 mm
Particle Size:
3 μm
Thermo ScientificTM HyPURITYTM C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
  • Highly characterized, tested and validated C18 columns
  • Ordering Information:
    Other column dimensions are also available. Please call Customer Service for more information.
    Specifications
    Column FormatAnalytical Column
    Diameter (Metric)4 mm
    For Use WithHPLC
    Length (Metric)150 mm
    Max. Pressure5800 psi (400 bar)
    Particle Size3 μm
    pH1 to 9
    Pore Size190 Å
    Surface Area200 m²/g
    Temperature60°C
    USP TypeL1
    Stationary PhaseC18
    Packing MaterialSpherical, Fully Porous, Base-Deactivated Silica
    Carbon Load13%
    Column TypeReversed Phase
    EndcappedYes
    PhaseReversed Phase
    Unit SizeEach