Replacement Filters for Unifilter Direct-Connection/ColumnPlus
Replacement Filters for Unifilter Direct-Connection/ColumnPlus
Actual product may vary
Replacement Filters for Unifilter Direct-Connection/ColumnPlus
Thermo Scientific™ Replacement Filters for Unifilter Direct-Connection/ColumnPlus are offered as particle size 0.5μm.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
22017
also known as AA22017
Replacement Filter
22155
also known as Q2215
Replacement Filter
Catalog number 22017
also known as AA22017
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Replacement Filter
Thermo Scientific™ Replacement Filters for Unifilter Direct-Connection/ColumnPlus are offered as particle size 0.5μm.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Diameter (Metric) Inner2.1, 3 mm
For Use With (Equipment)Unifilter Direct-Connection/Column
Certifications/ComplianceANSI-compliant polyester mesh
ColorOrange
Size2X-Large to 3X-Large
MaterialPolyester
DescriptionReplacement Filters; 0.5 U m; 2.1/3.0 mm
TypeReplacement Filter
Unit SizeEach