Reservoir Accessories
Actual product may vary
Reservoir Accessories
Level control and sterile overflow for storage reservoirs are perfect accessories for running reverse osmosis systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
22.0091Sterile Vent FilterStorage Tank
50135142Sterile Vent FilterStorage Tank 30 to 100 L
Catalog number 22.0091
HSN 84213920
Price (MYR)
911.00
Each
Type:
Sterile Vent Filter
For Use With (Equipment):
Storage Tank
Price (MYR)
911.00
Each
Level control and sterile overflow, Thermo Scientific™ Reservoir Accessories, are perfect for running reverse osmosis systems.
Level Control for Storage Reservoirs
  • Required accessory for running any reverse osmosis system; will be adapted to the specific reservoir during the order procedure
Sterile Overflow for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter for Storage Reservoirs
  • Recommended accessory to prevent organic contamination inside of reverse osmosis reservoirs
Vent filter with CO2 absorber for Storage Reservoirs
  • Recommended accessory to prevent organic and CO2 contamination inside of reverse osmosis reservoirs
Ordering Information:
This product is not available in all regions
Specifications
Description0.2 μm
For Use With (Equipment)Storage Tank
TypeSterile Vent Filter
Unit SizeEach