HyPURITY™ ADVANCE™ HPLC Columns, 5μm Particle Size
HyPURITY™ ADVANCE™ HPLC Columns, 5μm Particle Size
Actual product may vary
HyPURITY™ ADVANCE™ HPLC Columns, 5μm Particle Size
Use this specialty C8 column for separating compounds showing poor peak shape with traditional alkyl bonded phases.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric)
21005-154630150 mm4.6 mm
Catalog number 21005-154630
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
Diameter (Metric):
4.6 mm
Thermo Scientific™ HyPURITY ADVANCE™ HPLC columns combine the outstanding performance of our HyPURITY 190Å silica with a C8 reversed phase chemistry. This specialized packing with polar embedded character is well suited for LC/MS applications. The polar group protects the surface silanols from interaction with polar analytes and provides an alternate selectivity with excellent performance for compounds showing poor peak shape with traditional alkyl bonded phases such as C18.
HyPURITY ADVANCE HPLC columns offer:
  • A polar embedded stationary phase for alternate selectivity
  • Ideal for high throughput fast analyses
  • Enhanced aqueous stability
  • Exceptional peak shape for acids, bases and chelators
  • 3μm and 5μm particle sizes available in a wide range of column geometries
  • Suitable for LC/MS due to exceptionally low bleed
  • Ideally suited to a variety of pharmaceutical applications
Specifications
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Particle Size5 μm
Pore Size180 Å
Surface Area200 m²/g
USP TypeL7
Stationary PhaseC8
Packing MaterialSilica
Carbon Load9%
Column TypeReversed Phase
PhaseReversed Phase
Unit SizeEach