Internal Chamber Replacement Kit
Product Image
Internal Chamber Replacement Kit
For Use With: 310 Series (w/HEPA option) Incubators
Have Questions?
Catalog NumberFor Use With (Equipment)
190868310 Series Incubators w/HEPA Opt.
Catalog number 190868
Price (MYR)
-
For Use With (Equipment):
310 Series Incubators w/HEPA Opt.
Help ensure measurable Class 100 air quality performance with Thermo Scientific™ Replacement HEPA Filters and Cleaning Kits.
HEPA Filters
  • 99.97% filtering at 0.3μm
  • Compatible with all Thermo Scientific™ Forma™ Series II 3110, Thermo Scientific™ Steri-Cycle™, and Forma™ 310 Series (with HEPA filtration system option) incubators
  • Main chamber filter available as a stand-alone or packaged in a group with the other chamber in-line filters
Decontamination Kits
  • Consists of filters for difficult-to-clean chambers and gaskets (except door) and tubing
  • Reduces downtime and labor spent on decontamination
Specifications
DescriptionInternal chamber replacement kit with HEPA, gaskets, and other parts
For Use With (Equipment)310 Series Incubators w/HEPA Opt.
LightingLED
TypeInternal Chamber Replacement Kit
Unit SizeEA